Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016112.2 | NP_057196.2 | 1, 2 | 617 | ||
GenBank mRNA | AF053316.1 | - | 1, 2 | 109 | ||
AF073481.1 | - | 1, 2 | 617 | |||
BC025665.1 | - | 1, 2 | 314 | 5200440 |