Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 3 | 530 | ||
NM_001141932.1 | NP_001135404.1 | 3 | 530 | |||
NM_020296.2 | NP_064692.2 | 3 | 723 | |||
GenBank mRNA | AB026569.1 | - | 3 | 446 | ||
AK133054.1 | - | 3 | 251 | |||
AK135280.1 | - | 3 | 410 | |||
AK147109.1 | - | 3 | 724 | |||
AK161102.1 | - | 3 | 480 | |||
AK164307.1 | - | 3 | 524 | |||
AK184927.1 | - | 3 | 368 | |||
BC016501.1 | - | 3 | 711 | 4527742 | ||
BC057866.1 | - | 3 | 562 | 5327143 |