Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltb4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020490.2 | NP_065236.1 | 2 | 250 | ||
GenBank mRNA | AB029893.1 | - | 1 | 117 | ||
AK018029.1 | - | 2 | 251 | |||
AK036836.1 | - | 607 | ||||
BC106834.2 | - | 1 | 117 | 40044859 | ||
BC106835.2 | - | 1 | 117 | 40044860 |
Set Membership: |
G-protein coupled receptor |