Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290821.1 | NP_001277750.1 | 2 | 637 | ||
NM_020492.4 | NP_065238.2 | 2 | 637 | |||
GenBank mRNA | AK078258.1 | - | 2 | 637 | ||
AY129229.1 | - | 2 | 184 | |||
BC131949.1 | - | 2 | 302 | 40130226 | ||
BC137725.1 | - | 2 | 302 | 8860744 | ||
S73717.1 | - | 2 | 229 | |||
S73718.1 | - | 2 | 229 |