Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020497.2 | NP_065243.2 | 4 | 1396 | ||
GenBank mRNA | AF178935.1 | - | 3 | 438 | ||
AK028939.1 | - | 4 | 1349 | |||
AK043258.1 | - | 4 | 1321 | |||
AK133363.1 | - | 4 | 1397 | |||
BC048688.1 | - | 1 | 421 | |||
BC066056.1 | - | 4 | 1307 | |||
BC078415.1 | - | 4 | 812 | |||
BC138579.1 | - | 3 | 388 | 8861599 | ||
BC138580.1 | - | 3 | 388 | 8861600 | ||
BC145339.1 | - | 3 | 388 | |||
BC145340.1 | - | 3 | 388 |