Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164621.1 | NP_001158093.1 | 4 | 766 | ||
NM_001164622.1 | NP_001158094.1 | 3 | 453 | |||
NM_020012.2 | NP_064396.2 | 3 | 453 | |||
GenBank mRNA | AF249667.1 | - | 3 | 399 | ||
AF249668.1 | - | 2 | 136 | |||
AK009783.1 | - | 3 | 408 | |||
AK010162.1 | - | 3 | 410 | |||
AK011316.2 | - | 4 | 454 | |||
AK038986.1 | - | 3 | 395 | |||
AK180263.1 | - | 1 | 44 | |||
AK203098.1 | - | 44 | ||||
BC049079.1 | - | 4 | 766 | |||
BC054841.1 | - | 3 | 383 | 6489903 | ||
BC094250.1 | - | 3 | 404 | 5720753 | ||
CA315275.1 | - | 3 | 452 |