Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027044.3 | NP_081320.2 | 5 | 395 | ||
GenBank mRNA | AF108357.1 | - | 5 | 355 | ||
AK004438.1 | - | 5 | 350 | |||
AK005682.1 | - | 5 | 341 | |||
AK028206.1 | - | 5 | 332 | |||
AK145883.1 | - | 5 | 348 | |||
AK205037.1 | - | 1 | 50 | |||
BC026920.1 | - | 5 | 332 | 3666296 | ||
CN835522.1 | - | 5 | 398 |