Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020046.3 | NP_064430.1 | 3 | 723 | ||
GenBank mRNA | AF029667.2 | - | 3 | 428 | ||
AK030798.1 | - | 213 | ||||
AK088863.1 | - | 3 | 445 | |||
AK149350.1 | - | 3 | 471 | |||
BC019542.1 | - | 3 | 433 | 4039461 | ||
BC027829.1 | - | 3 | 684 | 5253723 | ||
BC045206.1 | - | 3 | 673 | 5253764 |