Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020259.4 | NP_064655.4 | 2, 3 | 973 | ||
GenBank mRNA | AF116865.1 | - | 2, 3 | 954 | ||
AK031539.1 | - | 2, 3 | 972 | |||
AK031841.1 | - | 2, 3 | 483 | |||
BC053012.1 | - | 2, 3 | 958 | 6402422 |