Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020265.4 | NP_064661.2 | 2 | 1035 | ||
GenBank mRNA | AJ243963.2 | - | 2 | 435 | ||
AK028225.1 | - | 2 | 320 | |||
AK031749.1 | - | 2 | 1036 | |||
BC096448.1 | - | 2 | 1026 | 6389874 |