Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146200.1 | NP_001139672.1 | 9, 10 | 3024 | ||
NM_001146201.1 | NP_001139673.1 | 9, 10 | 3080 | |||
NM_020272.2 | NP_064668.2 | 9, 10 | 3201 | |||
GenBank mRNA | AF208345.1 | - | 8, 9 | 2880 | ||
AJ249280.1 | - | 9, 10 | 3201 | |||
AK040543.1 | - | 9, 10 | 2986 | |||
AK077772.1 | - | 3, 4 | 356 | |||
AK132553.1 | - | 9, 10 | 3026 | |||
AK137470.1 | - | 9, 10 | 2933 | |||
AK137620.1 | - | 9, 10 | 3082 | |||
AK143238.1 | - | 6, 7 | 705 | |||
BC051246.1 | - | 9, 10 | 2941 | 6514979 |