Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039386.1 | NP_001034475.1 | 11, 12 | 1403 | ||
NM_001039387.1 | NP_001034476.1 | 10, 11 | 1397 | |||
NM_001177654.1 | NP_001171125.1 | 10, 11 | 1397 | |||
NM_001177655.1 | NP_001171126.1 | 9, 10 | 1307 | |||
NM_020276.3 | NP_064672.2 | 10, 11 | 1334 | |||
GenBank mRNA | AF266508.2 | - | 9, 10 | 1300 | ||
AJ278902.1 | - | 11, 12 | 1181 | |||
AK004986.2 | - | 10, 11 | 1394 | |||
AK019560.1 | - | 11, 12 | 1370 | |||
AK053402.1 | - | 9, 10 | 1254 | |||
AK158803.1 | - | 11, 12 | 1133 | |||
AK165300.1 | - | 10, 11 | 1342 | |||
BC006642.1 | - | 10, 11 | 1355 | 3483536 |