Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019098.4 | NP_061971.3 | 1 | 144 | ||
GenBank mRNA | AF272900.1 | - | 1 | 142 | ||
AL713036.1 | - | 1 | 145 | |||
BC150601.1 | - | 1 | 122 | 9056971 |