Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020506.1 | NP_065252.1 | 4 | 384 | ||
GenBank mRNA | AF145021.1 | - | 4 | 384 | ||
AK084907.1 | - | 5 | 569 | |||
AK129430.1 | - | 4 | 379 |