Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020516.2 | NP_065262.1 | 3 | 365 | ||
GenBank mRNA | AF019111.2 | - | 3 | 374 | ||
BC018216.1 | - | 3 | 365 | 4481698 |