Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020563.3 | NP_065588.1 | 2 | 117 | ||
GenBank mRNA | AF008595.1 | - | 2 | 118 | ||
BC115490.1 | - | 2 | 118 | 40100845 | ||
BC115491.1 | - | 2 | 118 | 40100846 | ||
GU269233.1 | - | 2 | 101 |