Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 8 | 1075 | ||
NM_001252490.1 | NP_001239419.1 | 7 | 949 | |||
NM_001252491.1 | NP_001239420.1 | 6 | 1155 | |||
NM_001252492.1 | NP_001239421.1 | 7 | 1318 | |||
NM_001252493.1 | NP_001239422.1 | 7 | 1318 | |||
NM_207530.3 | NP_997413.2 | 21 | 2204 | |||
GenBank mRNA | AB017026.1 | - | 5 | 1199 | ||
AF394063.2 | - | 5 | 510 | |||
AK050641.1 | - | 7 | 873 | |||
AK132676.1 | - | 8 | 1074 | |||
AK133806.1 | - | 5 | 581 | |||
AK168366.1 | - | 7 | 1279 | |||
AK200556.1 | - | 3 | 233 | |||
AY536214.1 | - | 21 | 2083 | |||
BB667982.1 | - | 3 | 277 | |||
BC015075.1 | - | 7 | 1039 | |||
BC046466.1 | - | 7 | 1081 | |||
BC057194.1 | - | 6 | 582 | |||
BC076637.1 | - | 7 | 949 | |||
BC092247.1 | - | 6 | 1155 | |||
CO426253.1 | - | 1 | 71 | |||
DV068702.1 | - | 5 | 545 |