Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020579.2 | NP_065604.2 | 6 | 1091 | ||
GenBank mRNA | AF142671.1 | - | 6 | 1031 | ||
AK155471.1 | - | 7 | 1191 | |||
BC004095.1 | - | 6 | 848 | |||
BC013619.1 | - | 6 | 1064 | 3965561 | ||
BC021830.1 | - | 4 | 1887 | |||
BC035546.1 | - | 1 | 886 |