Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020584.2 | NP_065609.2 | 1 | 600 | ||
GenBank mRNA | AB041557.1 | - | 1 | 554 | ||
AB041559.1 | - | 1 | 555 | |||
AK041613.1 | - | 1 | 546 | |||
AK047222.1 | - | 1 | 557 | |||
AK051818.1 | - | 1 | 520 | |||
AK053864.1 | - | 1 | 556 | |||
AK081557.1 | - | 1 | 556 | |||
AK084563.1 | - | 1 | 548 | |||
AK087729.1 | - | 1 | 558 | |||
AK148508.1 | - | 1 | 560 | |||
AK148537.1 | - | 1 | 562 | |||
AK161452.1 | - | 1 | 579 | |||
AK163044.1 | - | 1 | 556 | |||
AK165074.1 | - | 1 | 558 | |||
AK213108.1 | - | 1 | 106 | |||
BC012270.1 | - | 1 | 516 | 3483521 | ||
BC017641.1 | - | 1 | 576 | 4483320 |