Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem183a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042485.1 | NP_001035950.1 | 2, 3 | 332 | ||
NM_020588.2 | NP_065613.1 | 2, 3 | 332 | |||
GenBank mRNA | AB041592.1 | - | 2, 3 | 288 | ||
AK004922.2 | - | 2, 3 | 313 | |||
AK005019.1 | - | 2, 3 | 254 | |||
AK006183.1 | - | 2, 3 | 288 | |||
AK007779.1 | - | 2, 3 | 311 | |||
AK152852.1 | - | 2, 3 | 272 | |||
AK168331.1 | - | 2, 3 | 274 | |||
BC003210.1 | - | 2, 3 | 260 | 3156957 | ||
CF897336.1 | - | 2, 3 | 263 |