Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020590.4 | NP_065615.1 | 2 | 382 | ||
GenBank mRNA | AB041648.1 | - | 2 | 406 | ||
AF180518.1 | - | 2 | 409 | |||
AK006103.1 | - | 2 | 181 | |||
AK011975.1 | - | 2 | 377 | |||
AK014083.1 | - | 2 | 196 | |||
AK015049.1 | - | 2 | 199 | |||
AK018687.1 | - | 2 | 383 | |||
AK159015.1 | - | 2 | 377 | |||
AK167500.1 | - | 2 | 375 | |||
AK168921.1 | - | 2 | 414 | |||
AK182413.1 | - | 2 | 156 | |||
BC004602.1 | - | 2 | 317 | 3585132 | ||
BC024706.1 | - | 2 | 381 | 3711973 |