Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173483.1 | NP_001166954.1 | 4 | 569 | ||
NM_020599.2 | NP_065624.1 | 4 | 651 | |||
GenBank mRNA | AK014227.1 | - | 4 | 342 | ||
AK140622.1 | - | 4 | 570 | |||
BC016268.1 | - | 3 | 244 | 4481973 |