Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021278.2 | NP_067253.1 | 3 | 366 | ||
GenBank mRNA | AK010659.1 | - | 3 | 310 | ||
AK088783.1 | - | 3 | 310 | |||
AK162286.1 | - | 3 | 311 | |||
AK169108.1 | - | 3 | 313 | |||
AK169718.1 | - | 3 | 310 | |||
AK205579.1 | - | 1 | 91 | |||
BC018286.1 | - | 3 | 282 | 3501378 | ||
BQ127748.1 | - | 3 | 366 | |||
X16053.1 | - | 3 | 395 |