Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021284.6 | NP_067259.4 | 2, 3 | 313 | ||
GenBank mRNA | AK155064.1 | - | 2, 3 | 314 | ||
AK164065.1 | - | 2, 3 | 314 | |||
AY487807.1 | - | 1, 2 | 137 | |||
BC004642.1 | - | 2, 3 | 301 | 3158212 | ||
BC010202.1 | - | 2, 3 | 301 | 3158212 | ||
BC119495.1 | - | 3, 4 | 177 | 40052024 | ||
BC141051.1 | - | 1, 2 | 139 | 9055663 |