Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021137.4 | NP_066960.1 | 2 | 527 | ||
XM_017024993.1 | XP_016880482.1 | 2 | 483 | |||
GenBank mRNA | AK312387.1 | - | 2 | 288 | ||
BC001643.2 | - | 2 | 274 | 2823154 | ||
BC001949.1 | - | 2 | 274 | 2823154 | ||
BC003694.2 | - | 2 | 289 | |||
BT020121.1 | - | 1 | 78 | |||
CR456819.1 | - | 1 | 78 | |||
DB093553.1 | - | 2 | 527 | |||
M80783.1 | - | 2 | 231 |