Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020565.2 | NP_065590.2 | 3 | 397 | ||
GenBank mRNA | AF026075.1 | - | 3 | 398 | ||
BC125593.1 | - | 3 | 397 | 40130108 | ||
BC125595.1 | - | 3 | 397 | 40130110 |
Gene Symbol: Gm4794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101452.1 | NP_001094922.1 | 2 | 336 |