Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020567.2 | NP_065592.1 | 4 | 292 | ||
GenBank mRNA | AF068780.1 | - | 4 | 439 | ||
AK131959.1 | - | 4 | 266 | |||
BC020061.1 | - | 4 | 444 | 3988065 | ||
BY045588.1 | - | 4 | 293 |