Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020579.2 | NP_065604.2 | 8 | 1568 | ||
GenBank mRNA | AF142671.1 | - | 8 | 1508 | ||
AK155471.1 | - | 9 | 1668 | |||
BC004095.1 | - | 8 | 1325 | |||
BC013619.1 | - | 8 | 1541 | 3965561 | ||
BC021830.1 | - | 6 | 2364 | |||
BC035546.1 | - | 2 | 2859 | |||
CD363941.1 | - | 3 | 480 |