Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020580.3 | NP_065605.2 | 2 | 212 | ||
GenBank mRNA | AJ238741.1 | - | 2 | 146 | ||
AJ238742.1 | - | 2 | 212 | |||
AJ238743.1 | - | 2 | 166 | |||
AK010360.1 | - | 2 | 159 | |||
AK049418.1 | - | 2 | 184 | |||
AK159866.1 | - | 2 | 165 | |||
BC006959.1 | - | 2 | 173 | 3707929 | ||
BC031747.1 | - | 2 | 170 | 3673038 |