Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113527.1 | NP_001106999.1 | 3 | 809 | ||
NM_001291220.1 | NP_001278149.1 | 4 | 1127 | |||
NM_001291221.1 | NP_001278150.1 | 4 | 793 | |||
NM_020583.5 | NP_065608.2 | 4 | 1000 | |||
GenBank mRNA | AK005528.1 | - | 4 | 794 | ||
AK008139.1 | - | 3 | 810 | |||
AK008171.1 | - | 3 | 810 | |||
AK008353.1 | - | 3 | 812 | |||
AK136756.1 | - | 4 | 926 | |||
BC022751.1 | - | 4 | 947 | 4912320 |