Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020584.2 | NP_065609.2 | 2 | 827 | ||
GenBank mRNA | AB041557.1 | - | 2 | 781 | ||
AB041559.1 | - | 2 | 782 | |||
AK041613.1 | - | 2 | 773 | |||
AK051818.1 | - | 2 | 747 | |||
AK081557.1 | - | 2 | 783 | |||
AK084563.1 | - | 2 | 775 | |||
AK087729.1 | - | 2 | 785 | |||
AK148508.1 | - | 2 | 787 | |||
AK148537.1 | - | 2 | 789 | |||
AK161452.1 | - | 2 | 806 | |||
AK165074.1 | - | 2 | 785 | |||
AK213108.1 | - | 2 | 334 | |||
BC012270.1 | - | 2 | 743 | 3483521 | ||
BC017641.1 | - | 2 | 803 | 4483320 |