Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021274.2 | NP_067249.1 | 4 | 370 | ||
GenBank mRNA | AK144279.1 | - | 4 | 361 | ||
AK150380.1 | - | 4 | 361 | |||
AK150765.1 | - | 4 | 361 | |||
AK151210.1 | - | 4 | 361 | |||
AK151248.1 | - | 4 | 361 | |||
AK151415.1 | - | 4 | 361 | |||
AK151534.1 | - | 4 | 361 | |||
AK152568.1 | - | 4 | 361 | |||
AK152814.1 | - | 4 | 361 | |||
AK152924.1 | - | 3 | 203 | |||
AK153181.1 | - | 4 | 361 | |||
AK157130.1 | - | 4 | 361 | |||
AK157139.1 | - | 4 | 361 | |||
AK157589.1 | - | 4 | 361 | |||
AK157678.1 | - | 4 | 361 | |||
AK172540.1 | - | 4 | 361 | |||
BC030067.1 | - | 4 | 337 | 1446589 | ||
M33266.1 | - | 4 | 335 |