Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267695.2 | NP_001254624.2 | 4, 5 | 536 | ||
NM_021281.3 | NP_067256.4 | 4, 5 | 533 | |||
GenBank mRNA | AF038546.1 | - | 4, 5 | 475 | ||
AJ002386.1 | - | 4, 5 | 488 | |||
AK028366.1 | - | 4, 5 | 535 | |||
AK150274.1 | - | 4, 5 | 467 | |||
AK152191.1 | - | 4, 5 | 486 | |||
AK153540.1 | - | 1, 2 | 135 | |||
BC002125.1 | - | 4, 5 | 442 | 3495719 |