Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021284.6 | NP_067259.4 | 3 | 324 | ||
GenBank mRNA | AK155064.1 | - | 3 | 325 | ||
AK164065.1 | - | 3 | 325 | |||
AY487807.1 | - | 2 | 148 | |||
BC004642.1 | - | 3 | 312 | 3158212 | ||
BC010202.1 | - | 3 | 312 | 3158212 | ||
BC119495.1 | - | 4 | 188 | 40052024 | ||
BC141051.1 | - | 2 | 150 | 9055663 |