Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020575.2 | NP_065600.1 | 5 | 1279 | ||
GenBank mRNA | AF155739.1 | - | 5 | 1279 | ||
AK034109.1 | - | 1269 | ||||
AK143177.1 | - | 5 | 1301 | |||
AK166666.1 | - | 5 | 1286 | |||
AK167246.1 | - | 5 | 1261 | |||
AK168555.1 | - | 5 | 1282 | |||
BC025029.1 | - | 5 | 1243 | 4935124 |