Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020577.2 | NP_065602.2 | 9 | 957 | ||
GenBank mRNA | AF166383.1 | - | 9 | 927 | ||
AK002801.1 | - | 9 | 888 | |||
AK006571.1 | - | 9 | 942 | |||
AK008104.1 | - | 1 | 61 | |||
AK009814.1 | - | 8 | 922 | |||
BC013468.1 | - | 9 | 882 | |||
BC145663.1 | - | 9 | 899 | 40130966 | ||
BQ445127.1 | - | 2 | 175 |