Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042484.1 | NP_001035949.1 | 6 | 1078 | ||
NM_020585.2 | NP_065610.2 | 5 | 1291 | |||
GenBank mRNA | AB041568.1 | - | 5 | 1227 | ||
AK048175.1 | - | 6 | 1054 | |||
AK077582.1 | - | 6 | 1052 | |||
AK150462.1 | - | 6 | 1072 | |||
AK150696.1 | - | 5 | 1233 | |||
AK151353.1 | - | 6 | 1072 | |||
AK152520.1 | - | 6 | 1072 | |||
AK152606.1 | - | 6 | 1051 | |||
AK153213.1 | - | 6 | 1072 | |||
AK158227.1 | - | 5 | 1197 | |||
AK170129.1 | - | 6 | 1057 | |||
AK211526.1 | - | 1 | 108 | |||
AK212515.1 | - | 1 | 108 | |||
BC016894.1 | - | 6 | 1030 | 4223937 | ||
BC019697.1 | - | 5 | 1216 | |||
BC058523.1 | - | 5 | 1134 | 5717934 | ||
BC093518.1 | - | 6 | 1048 | 3157520 | ||
BC103796.1 | - | 6 | 1043 | 6519018 |