Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020603.2 | NP_065628.1 | 14 | 1723 | ||
GenBank mRNA | AK009209.1 | - | 2 | 139 | ||
AK145068.1 | - | 14 | 1778 | |||
AK178374.1 | - | 3 | 278 | |||
AK186164.1 | - | 3 | 276 | |||
BC013527.1 | - | 14 | 1766 | |||
BC026431.1 | - | 11 | 1347 | |||
BC046977.1 | - | 14 | 1723 | 6315172 | ||
KJ650211.1 | - | 14 | 1708 | |||
KJ663716.1 | - | 14 | 1576 |