Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021050.2 | NP_066388.1 | 14 | 1977 | ||
GenBank mRNA | M60493.1 | - | 14 | 1978 | ||
M69298.1 | - | 14 | 1976 | |||
X72693.1 | - | 4 | 306 | |||
X72694.1 | - | 4 | 305 |