Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013437.1 | NP_001013455.1 | 1, 2 | 257 | ||
NM_031216.3 | NP_112493.2 | 1, 2 | 257 | |||
XM_005258152.4 | XP_005258209.1 | 1, 2 | 247 | |||
XM_011525742.2 | XP_011524044.1 | 1, 2 | 253 | |||
XM_011525743.2 | XP_011524045.1 | 1, 2 | 247 | |||
XM_011525744.1 | XP_011524046.1 | 1, 2 | 253 | |||
XM_017026025.1 | XP_016881514.1 | 1, 2 | 253 | |||
GenBank mRNA | AF136976.1 | - | 1, 2 | 225 | ||
AF255625.1 | - | 1, 2 | 161 | |||
AF431970.1 | - | 1, 2 | 161 | |||
AF514996.1 | - | 1, 2 | 121 | |||
AK056940.1 | - | 1, 2 | 220 | |||
AK291226.1 | - | 1, 2 | 220 | |||
AK314403.1 | - | 1, 2 | 231 | |||
BC012430.1 | - | 1, 2 | 211 | 3865994 | ||
CB990796.1 | - | 284 |