Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111058.1 | NP_001104528.1 | 2 | 422 | ||
NM_021293.3 | NP_067268.1 | 2 | 422 | |||
GenBank mRNA | AK046303.1 | - | 4 | 645 | ||
AK089205.1 | - | 2 | 339 | |||
AK155745.1 | - | 2 | 351 | |||
AK170937.1 | - | 2 | 390 | |||
BC132379.1 | - | 2 | 422 | 40130656 | ||
S71345.1 | - | 2 | 353 | |||
S71403.1 | - | 2 | 353 |