Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021296.2 | NP_067271.1 | 4 | 1560 | ||
GenBank mRNA | AK012634.1 | - | 4 | 1539 | ||
AK082983.1 | - | 1551 | ||||
AK145493.1 | - | 4 | 1552 | |||
BC068232.1 | - | 4 | 1537 | 30533079 | ||
BC094583.1 | - | 4 | 1527 | |||
BC120747.1 | - | 4 | 1550 | 40129670 |