Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317399.1 | NP_001304328.1 | 3, 4 | 665 | ||
NM_031954.4 | NP_114160.1 | 3, 4 | 665 | |||
NR_133898.1 | 2, 3 | 451 | ||||
GenBank mRNA | AF113208.1 | - | 3, 4 | 410 | ||
AK027543.1 | - | 3, 4 | 471 | |||
AK222548.1 | - | 3, 4 | 421 | |||
AK309783.1 | - | 2, 3 | 280 | |||
AY205299.1 | - | 3, 4 | 471 | |||
AY597809.1 | - | 2, 3 | 383 | |||
BC040062.1 | - | 3, 4 | 418 | 5494016 | ||
CX785497.1 | - | 2, 3 | 194 |