Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021320.3 | NP_067295.2 | 2 | 743 | ||
GenBank mRNA | AF268066.1 | - | 2 | 338 | ||
AF281278.1 | - | 2 | 591 | |||
BB609851.1 | - | 2 | 614 | |||
BC119182.1 | - | 2 | 425 | 8733931 | ||
BC119184.1 | - | 2 | 425 | 8733933 |