Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021339.2 | NP_067314.2 | 3, 4 | 530 | ||
GenBank mRNA | AF090866.1 | - | 3, 4 | 530 | ||
AK040711.1 | - | 4, 5 | 622 | |||
AK157750.1 | - | 5, 6 | 713 | |||
BC109176.2 | - | 3, 4 | 529 | 40048829 |