Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159650.1 | NP_001153122.1 | 2, 3 | 138 | ||
NM_021352.3 | NP_067327.1 | 2, 3 | 208 | |||
GenBank mRNA | AJ272229.2 | - | 1, 2 | 192 | ||
BC031442.1 | - | 1, 2 | 199 | 4501690 | ||
BC058522.1 | - | 1, 2 | 117 | 6824229 | ||
CJ046471.1 | - | 2, 3 | 209 |