Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163480.1 | NP_001156952.1 | 2, 3 | 497 | ||
NM_021360.4 | NP_067335.4 | 2, 3 | 975 | |||
GenBank mRNA | AF400063.1 | - | 2, 3 | 546 | ||
AF401228.1 | - | 2, 3 | 432 | |||
AK010787.1 | - | 2, 3 | 644 | |||
AK046382.1 | - | 2, 3 | 774 | |||
BC058386.1 | - | 2, 3 | 800 | 6831952 | ||
BC099702.1 | - | 2, 3 | 975 | 6825751 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | Y15160.1 | - | 328 | Chr. - - [-] on Build |