Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB162929.1 | - | 10 | 1067 | ||
AF190699.1 | - | 10 | 1139 | |||
AF190991.1 | - | 10 | 1139 | |||
AK015486.1 | - | 9 | 790 | |||
AK018148.1 | - | 10 | 1131 | |||
AK031550.1 | - | 10 | 1167 | |||
AK042758.1 | - | 10 | 920 | |||
AK043998.1 | - | 10 | 1187 | |||
AK048413.1 | - | 10 | 1138 | |||
AK049903.1 | - | 9 | 873 | |||
AK081981.1 | - | 10 | 1141 | |||
AK083126.1 | - | 9 | 1264 | |||
BC046310.1 | - | 10 | 1125 | 6307420 | ||
BC065694.1 | - | 10 | 1131 | 6813365 |