Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163514.1 | NP_001156986.1 | 3 | 643 | ||
NM_001163515.1 | NP_001156987.1 | 2 | 761 | |||
NM_021388.4 | NP_067363.3 | 3 | 836 | |||
GenBank mRNA | AB032170.1 | - | 3 | 646 | ||
AB032171.1 | - | 3 | 631 | |||
AF200973.1 | - | 3 | 608 | |||
AK019370.1 | - | 3 | 574 | |||
AK028677.1 | - | 3 | 809 | |||
AK032016.1 | - | 1 | 169 | |||
AK159281.1 | - | 3 | 823 | |||
AK161866.1 | - | 2 | 727 | |||
BC031438.1 | - | 3 | 800 | 4024555 | ||
BC094444.1 | - | 3 | 549 | 4507232 |