Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139126.3 | NP_624311.1 | 2, 3 | 245 | ||
GenBank mRNA | AF357880.1 | - | 2, 3 | 178 | ||
AK092069.1 | - | 2, 3 | 178 | |||
AK315388.1 | - | 2, 3 | 245 | |||
BC020986.1 | - | 2, 3 | 181 | 3852013 | ||
BC064134.1 | - | 2, 3 | 163 | |||
BC107684.1 | - | 2, 3 | 179 | |||
BX537536.1 | - | 2, 3 | 179 |